missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4-methylpyridine 98.0+%, TCI America™

$112.17 - $345.64

Chemical Identifiers

CAS 4926-28-7
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00082590
InChI Key LSZMVESSGLHDJE-UHFFFAOYSA-N
Synonym 2 bromo-4-methyl pyridine
PubChem CID 2734087
IUPAC Name 2-bromo-4-methylpyridine
SMILES CC1=CC(=NC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21125G View Documents Tci America
B21125G
5g Each for $112.17
 
B211225G View Documents Tci America
B211225G
25g Each for $345.64
 
Specifications

Chemical Identifiers

4926-28-7
172.025
LSZMVESSGLHDJE-UHFFFAOYSA-N
2734087
CC1=CC(=NC=C1)Br
C6H6BrN
MFCD00082590
2 bromo-4-methyl pyridine
2-bromo-4-methylpyridine

Specifications

Yellow
4926-28-7
MFCD00082590
LSZMVESSGLHDJE-UHFFFAOYSA-N
2-bromo-4-methylpyridine
2734087
Clear Liquid at 20°C
2-Bromo-4-methylpyridine
5g
C6H6BrN
2 bromo-4-methyl pyridine
CC1=CC(=NC=C1)Br
172.025
172.03
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No