Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-methylpyridine, 96%

$126.57 - $518.32

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-methylpyridine
CAS 4926-28-7
Molecular Formula C6H6BrN
Formula Weight 172.03
MDL Number MFCD00082590
Beilstein 107331
Synonym 2-Bromo-4-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2525306

  SDS
alfa aesar™
B2525306
5g Each for $126.57

AAB2525314

  SDS
alfa aesar™
B2525314
25g Each for $518.32
Description & Specifications

Specifications

Formula Weight 172.03
Boiling Point 87°C (10mmHg)
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00082590
Flash Point >110°C (230°F)
Molecular Formula C6H6BrN
Assay 96%
CAS 4926-28-7
Synonym 2-Bromo-4-picoline
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2-Bromo-4-methylpyridine
Beilstein 107331
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.