missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4'-methylpropiophenone, 98%

Manufacturer:  Alfa Aesar™ H6488106

 View more versions of this product

Catalog No. AAH6488106



Description

Description

Applications
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

Solubility
Soluble in chloroform and methanol.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

5g
C10H11BrO
2207628
Soluble in chloroform and methanol.
CC1=CC=C(C=C1)C(=O)C(C)Br
227.101
227.1
98%
1451-82-7
MFCD11131402
1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-1-4-methylphenyl-1-propanone, 2-bromo-1-p-tolyl propan-1-one, 2-bromo-1-p-tolyl-propan-1-one, 2-bromo-1-p-tolylpropan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-4-methylpropiophenone, 4'-methyl-alpha-bromopropiophenone, ksc917e0p
OZLUPIIIHOOPNQ-UHFFFAOYSA-N
2-bromo-1-(4-methylphenyl)propan-1-one
2734063
Solid

Chemical Identifiers

1451-82-7
227.101
OZLUPIIIHOOPNQ-UHFFFAOYSA-N
2734063
CC1=CC=C(C=C1)C(=O)C(C)Br
C10H11BrO
MFCD11131402
1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-1-4-methylphenyl-1-propanone, 2-bromo-1-p-tolyl propan-1-one, 2-bromo-1-p-tolyl-propan-1-one, 2-bromo-1-p-tolylpropan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-4-methylpropiophenone, 4'-methyl-alpha-bromopropiophenone, ksc917e0p
2-bromo-1-(4-methylphenyl)propan-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents