Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-methylphenyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-methylphenyl isothiocyanate
CAS 19241-39-5
Molecular Formula C8H6BrNS
Formula Weight 228.12
MDL Number MFCD00041040
UN Number UN2922
Beilstein 2717454
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0941303

  SDS
alfa aesar™
L0941303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0941306

  SDS
alfa aesar™
L0941306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 228.12
CAS 19241-39-5
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Chemical Name or Material 2-Bromo-4-methylphenyl isothiocyanate
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Molecular Formula C8H6BrNS
Quantity 1g
MDL Number MFCD00041040
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
UN Number UN2922
Boiling Point 119° to 120°C (0.5mmHg)
Odor Irritating
Beilstein 2717454
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.