Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-methylbenzaldehyde, 95%

$86.51 - $86.51

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-methylbenzaldehyde
CAS 824-54-4
Molecular Formula C8H7BrO
Formula Weight 199.05
MDL Number MFCD04039890
Synonym 2-Bromo-p-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2755803

 
alfa aesar™
H2755803
1g Each for $86.51
Description & Specifications

Specifications

Formula Weight 199.05
Chemical Name or Material 2-Bromo-4-methylbenzaldehyde
Assay 95%
Molecular Formula C8H7BrO
MDL Number MFCD04039890
CAS 824-54-4
Melting Point 33° to 37°C
Synonym 2-Bromo-p-tolualdehyde
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.