missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4-methylaniline 98%, ACROS Organics™

Manufacturer:  Acros Organics 154240100

 View more versions of this product

Catalog No. AC154240100



Specifications

Specifications

2-Bromo-4-methylaniline
Glass bottle
Yellow to Brown
98%
97.5
98%
BrC6H3(CH3)NH2
12, 991
UVRRJILIXQAAFK-UHFFFAOYSA-N
2-bromo-4-methylaniline
11422
Liquid
Authentic
1.6010 to 1.6030 (20°C, 589nm)
10mL
583-68-6
100.0
C7H8BrN
MFCD00007635
2-bromo-p-toluidine, benzenamine, 2-bromo-4-methyl, 4-amino-3-bromotoluene, 2-bromo-4-methylbenzenamine, p-toluidine, 2-bromo, 2-bromo-4-methyl-phenylamine, 3-bromo-4-aminotoluene, 4-methyl-2-bromoaniline, 2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline
CC1=CC(=C(C=C1)N)Br
186.052
186.05
≥97.5%

Chemical Identifiers

583-68-6
186.052
UVRRJILIXQAAFK-UHFFFAOYSA-N
11422
CC1=CC(=C(C=C1)N)Br
C7H8BrN
MFCD00007635
2-bromo-p-toluidine, benzenamine, 2-bromo-4-methyl, 4-amino-3-bromotoluene, 2-bromo-4-methylbenzenamine, p-toluidine, 2-bromo, 2-bromo-4-methyl-phenylamine, 3-bromo-4-aminotoluene, 4-methyl-2-bromoaniline, 2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline
2-bromo-4-methylaniline
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-515-7

RTECSNumber : XU4300000

TSCA : TSCA

SDS Documents