Learn More
2-Bromo-4-methylaniline 98%, ACROS Organics™
$68.08 - $68.08
Chemical Identifiers
CAS | 583-68-6 |
---|---|
Molecular Formula | C7H8BrN |
Molecular Weight (g/mol) | 186.052 |
MDL Number | MFCD00007635 |
InChI Key | UVRRJILIXQAAFK-UHFFFAOYSA-N |
Synonym | 2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline, 2-bromo-4-methyl-phenylamine, 2-bromo-4-methylbenzenamine, 2-bromo-p-toluidine, 3-bromo-4-aminotoluene, 4-amino-3-bromotoluene, 4-methyl-2-bromoaniline, benzenamine, 2-bromo-4-methyl, p-toluidine, 2-bromo |
PubChem CID | 11422 |
IUPAC Name | 2-bromo-4-methylaniline |
SMILES | CC1=CC(=C(C=C1)N)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC154240100
|
Thermo Fisher Scientific
154240100 |
10mL | Glass bottle |
Each for $68.08
|
|
Chemical Identifiers
583-68-6 | |
186.052 | |
UVRRJILIXQAAFK-UHFFFAOYSA-N | |
11422 | |
CC1=CC(=C(C=C1)N)Br |
C7H8BrN | |
MFCD00007635 | |
2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline, 2-bromo-4-methyl-phenylamine, 2-bromo-4-methylbenzenamine, 2-bromo-p-toluidine, 3-bromo-4-aminotoluene, 4-amino-3-bromotoluene, 4-methyl-2-bromoaniline, benzenamine, 2-bromo-4-methyl, p-toluidine, 2-bromo | |
2-bromo-4-methylaniline |
Specifications
Authentic | |
1.6010 to 1.6030 (20°C, 589nm) | |
10mL | |
97.5 | |
98% | |
BrC6H3(CH3)NH2 | |
12, 991 | |
UVRRJILIXQAAFK-UHFFFAOYSA-N | |
2-bromo-4-methylaniline | |
11422 | |
Liquid | |
2-Bromo-4-methylaniline, 98% |
Glass bottle | |
Yellow to Brown | |
583-68-6 | |
100.0 | |
C7H8BrN | |
MFCD00007635 | |
2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline, 2-bromo-4-methyl-phenylamine, 2-bromo-4-methylbenzenamine, 2-bromo-p-toluidine, 3-bromo-4-aminotoluene, 4-amino-3-bromotoluene, 4-methyl-2-bromoaniline, benzenamine, 2-bromo-4-methyl, p-toluidine, 2-bromo | |
CC1=CC(=C(C=C1)N)Br | |
186.052 | |
186.05 | |
≥97.5% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-515-7
RTECSNumber : XU4300000
TSCA : TSCA