Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Bromo-4'-methylacetanilide, 98%

$35.41 - $35.41

Chemical Identifiers

Chemical Name or Material 2'-Bromo-4'-methylacetanilide
CAS 614-83-5
Molecular Formula C9H10BrNO
Formula Weight 228.09
MDL Number MFCD00017781
Beilstein 2691019
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2221306

  SDS
alfa aesar™
B2221306
5g Each for $35.41
Description & Specifications

Specifications

Formula Weight 228.09
Chemical Name or Material 2'-Bromo-4'-methylacetanilide
TSCA TSCA
Assay 98%
Molecular Formula C9H10BrNO
MDL Number MFCD00017781
CAS 614-83-5
Melting Point 117° to 119°C
Beilstein 2691019
Quantity 5g