missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4-methyl-5-nitropyridine, TCI America™

$135.97 - $406.39

Chemical Identifiers

CAS 23056-47-5
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD03095066
InChI Key OSYUAHGEDKTDOX-UHFFFAOYSA-N
Synonym 2-Bromo-5-nitro-4-picoline
PubChem CID 345873
IUPAC Name 2-bromo-4-methyl-5-nitropyridine
SMILES CC1=CC(=NC=C1[N+](=O)[O-])Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B50625G View Documents TCI America
B50625G
5g Each for $135.97
 
B506225G View Documents TCI America
B506225G
25g Each for $406.39
 
Specifications

Chemical Identifiers

23056-47-5
217.022
OSYUAHGEDKTDOX-UHFFFAOYSA-N
345873
CC1=CC(=NC=C1[N+](=O)[O-])Br
C6H5BrN2O2
MFCD03095066
2-Bromo-5-nitro-4-picoline
2-bromo-4-methyl-5-nitropyridine

Specifications

Yellow
5g
C6H5BrN2O2
2-Bromo-5-nitro-4-picoline
CC1=CC(=NC=C1[N+](=O)[O-])Br
217.022
217.02
≥98.0% (GC)
65°C
23056-47-5
MFCD03095066
OSYUAHGEDKTDOX-UHFFFAOYSA-N
2-bromo-4-methyl-5-nitropyridine
345873
Crystal-Powder at 20°C
2-Bromo-4-methyl-5-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No