Learn More
2-Bromo-4-isopropylaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1499514
Description
2-Bromo-4-isopropylaniline is involved in the preparation of 4-(5-aryl-1, 2, 3, 6-tetrahydropyridino) pyrimidine derivatives and 4-substituted-2-anilinopyrimidine derivatives, which find application as corticotropin releasing factor (CRF) antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4-isopropylaniline is involved in the preparation of 4-(5-aryl-1, 2, 3, 6-tetrahydropyridino) pyrimidine derivatives and 4-substituted-2-anilinopyrimidine derivatives, which find application as corticotropin releasing factor (CRF) antagonists.
Solubility
Miscible with dioxane and ethoxyethanol. Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
2-Bromo-4-isopropylaniline | |
1.35 | |
C9H12BrN | |
25 g | |
2638470 | |
Miscible with dioxane and ethoxyethanol. Immiscible with water. | |
CC(C)C1=CC=C(N)C(Br)=C1 | |
214.11 | |
214.11 |
51605-97-1 | |
116°C to 118°C (3 mmHg) | |
MFCD00051585 | |
UN2810 | |
2-bromo-4-isopropylaniline, 2-bromo-4-isopropyl-phenylamine, 4-amino-2-bromocumene, benzenamine, 2-bromo-4-1-methylethyl, zlchem 981, pubchem17446, acmc-1anzo, 2-bromo-4-isopropyl aniline, 2-bromo4-isopropyl-phenylamine, 2-bromo-4-2-propyl aniline | |
WEMDUNBELVTSRP-UHFFFAOYSA-N | |
2-bromo-4-(propan-2-yl)aniline | |
521337 | |
97% |
Chemical Identifiers
51605-97-1 | |
214.11 | |
WEMDUNBELVTSRP-UHFFFAOYSA-N | |
521337 |
C9H12BrN | |
MFCD00051585 | |
2-bromo-4-isopropylaniline, 2-bromo-4-isopropyl-phenylamine, 4-amino-2-bromocumene, benzenamine, 2-bromo-4-1-methylethyl, zlchem 981, pubchem17446, acmc-1anzo, 2-bromo-4-isopropyl aniline, 2-bromo4-isopropyl-phenylamine, 2-bromo-4-2-propyl aniline | |
CC(C)C1=CC=C(N)C(Br)=C1 |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only