missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-isopropylaniline, 97%

Manufacturer:  Alfa Aesar™ A1499522

 View more versions of this product

Catalog No. AAA1499522



Description

Description

Applications
2-Bromo-4-isopropylaniline is involved in the preparation of 4-(5-aryl-1, 2, 3, 6-tetrahydropyridino) pyrimidine derivatives and 4-substituted-2-anilinopyrimidine derivatives, which find application as corticotropin releasing factor (CRF) antagonists.

Solubility
Miscible with dioxane and ethoxyethanol. Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

2-Bromo-4-isopropylaniline
100g
C9H12BrN
UN2810
2-bromo-4-isopropylaniline, 2-bromo-4-isopropyl-phenylamine, 4-amino-2-bromocumene, benzenamine, 2-bromo-4-1-methylethyl, zlchem 981, pubchem17446, acmc-1anzo, 2-bromo-4-isopropyl aniline, 2-bromo4-isopropyl-phenylamine, 2-bromo-4-2-propyl aniline
CC(C)C1=CC(=C(C=C1)N)Br
214.106
214.11
116°C to 118°C (3mmHg)
51605-97-1
MFCD00051585
2638470
WEMDUNBELVTSRP-UHFFFAOYSA-N
2-bromo-4-propan-2-ylaniline
521337
97%

Chemical Identifiers

51605-97-1
214.106
WEMDUNBELVTSRP-UHFFFAOYSA-N
521337
CC(C)C1=CC(=C(C=C1)N)Br
C9H12BrN
MFCD00051585
2-bromo-4-isopropylaniline, 2-bromo-4-isopropyl-phenylamine, 4-amino-2-bromocumene, benzenamine, 2-bromo-4-1-methylethyl, zlchem 981, pubchem17446, acmc-1anzo, 2-bromo-4-isopropyl aniline, 2-bromo4-isopropyl-phenylamine, 2-bromo-4-2-propyl aniline
2-bromo-4-propan-2-ylaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Documents