Please login to your online account to display your discounted pricing

2-Bromo-4-fluorophenol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-fluorophenol
Name Note 98%
CAS 496-69-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4BrFO
Linear Formula BrC6H3(F)OH
Formula Weight 191.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311070010

  SDS
thermo fisher scientific
AC311070010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Solid
Formula Weight 191.01
Quantity 1g
Color Light brown to brown
TSCA TSCA
Density 1.744g/mL
Melting Point 42° to 46°C
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H4BrFO
Linear Formula BrC6H3(F)OH
MDL Number MFCD00010614
CAS 496-69-5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Bromo-4-fluorophenol
Packaging Glass bottle
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.