missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluorobenzylamine hydrochloride, 96%

Manufacturer:  Alfa Aesar™ H62200MD

 View more versions of this product

Catalog No. AAH62200MD



Description

Description

Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

2-Bromo-4-fluorobenzylamine hydrochloride
289038-14-8
MFCD00672922
PBCSNHSXUSMNOV-UHFFFAOYSA-N
(2-bromo-4-fluorophenyl)methanamine;hydrochloride
2773353
Solid
250mg
C7H8BrClFN
1-2-bromo-4-fluorophenyl methanamine hydrochloride
C1=CC(=C(C=C1F)Br)CN.Cl
240.5
240.5
96%

Chemical Identifiers

289038-14-8
240.5
PBCSNHSXUSMNOV-UHFFFAOYSA-N
2773353
C1=CC(=C(C=C1F)Br)CN.Cl
C7H8BrClFN
MFCD00672922
1-2-bromo-4-fluorophenyl methanamine hydrochloride
(2-bromo-4-fluorophenyl)methanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents