missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4-fluorobenzyl bromide, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6215706

 View more versions of this product

Catalog No. AAH6215706



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Bromo-4-fluorobenzyl bromide
C7H5Br2F
5 g
2-bromo-4-fluorobenzyl bromide, 2-bromo-4-fluorobenzylbromide, 2-bromo-1-bromomethyl-4-fluorobenzene, benzene, 2-bromo-1-bromomethyl-4-fluoro, 4-fluoro-2-bromobenzylbromide, 1-bromo-2-bromomethyl-5-fluorobenzene, pubchem4883, 2-bromo-1-bromomethyl-4-fluoro-benzene, acmc-1b995
FC1=CC(Br)=C(CBr)C=C1
267.92
267.92
Solid
61150-57-0
MFCD03840524
UN3261
QPLUIZXBWYUFMY-UHFFFAOYSA-N
2-bromo-1-(bromomethyl)-4-fluorobenzene
10706977
96%

Chemical Identifiers

61150-57-0
267.92
QPLUIZXBWYUFMY-UHFFFAOYSA-N
10706977
FC1=CC(Br)=C(CBr)C=C1
C7H5Br2F
MFCD03840524
2-bromo-4-fluorobenzyl bromide, 2-bromo-4-fluorobenzylbromide, 2-bromo-1-bromomethyl-4-fluorobenzene, benzene, 2-bromo-1-bromomethyl-4-fluoro, 4-fluoro-2-bromobenzylbromide, 1-bromo-2-bromomethyl-5-fluorobenzene, pubchem4883, 2-bromo-1-bromomethyl-4-fluoro-benzene, acmc-1b995
2-bromo-1-(bromomethyl)-4-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions