Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluorobenzyl alcohol, 98%

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-fluorobenzyl alcohol
CAS 229027-89-8
Formula Weight 205.02
MDL Number MFCD00672925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6185014

  SDS
alfa aesar™
H6185014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6185022

  SDS
alfa aesar™
H6185022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 68° to 72°C
Formula Weight 205.02
Chemical Name or Material 2-Bromo-4-fluorobenzyl alcohol
Color Off-white
Physical Form Solid
Assay 98%
Quantity 25g
MDL Number MFCD00672925
CAS 229027-89-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Bromo-4-fluorobenzyl alcohol, is used as an alcohol, medicine intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.