missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluorobenzyl alcohol, 98%

Manufacturer:  Alfa Aesar™ H6185022

 View more versions of this product

Catalog No. AAH6185022



Description

Description

Applications
2-Bromo-4-fluorobenzyl alcohol, is used as an alcohol, medicine intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Bromo-4-fluorobenzyl alcohol
68°C to 72°C
229027-89-8
MFCD00672925
Soluble in water.
C1=CC(=C(C=C1F)Br)CO
205.026
205.02
98%
White
100g
C7H6BrFO
2-bromo-4-fluorobenzenemethanol
XGAMLBPEVCLQEJ-UHFFFAOYSA-N
(2-bromo-4-fluorophenyl)methanol
2773347
Solid

Chemical Identifiers

229027-89-8
205.026
XGAMLBPEVCLQEJ-UHFFFAOYSA-N
2773347
C1=CC(=C(C=C1F)Br)CO
C7H6BrFO
MFCD00672925
2-bromo-4-fluorobenzenemethanol
(2-bromo-4-fluorophenyl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents