Please login to your online account to display your discounted pricing

2-Bromo-4-fluorobenzotrifluoride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-fluorobenzotrifluoride
CAS 351003-21-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H3BrF4
Linear Formula BrC6H3(F)CF3
Formula Weight 243
MDL Number MFCD03094041
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378170010

  SDS
thermo fisher scientific
AC378170010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 243
Quantity 1g
Color Clear colorless to light yellow
Density 1.7530g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.4645 to 1.4665 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03094041
Chemical Name or Material 2-Bromo-4-fluorobenzotrifluoride
Molecular Formula C7H3BrF4
CAS Min % 97.5
CAS 351003-21-9
Linear Formula BrC6H3(F)CF3
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.