missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-4-fluorobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America B28875G
Specifications
2-Bromo-4-fluorobenzoic Acid | |
174°C | |
C7H4BrFO2 | |
5 g | |
RRKPMLZRLKTDQV-UHFFFAOYSA-N | |
2-bromo-4-fluorobenzoic acid | |
1268169 | |
≥98.0% (GC,T) |
1006-41-3 | |
White | |
MFCD00055370 | |
2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid | |
C1=CC(=C(C=C1F)Br)C(=O)O | |
219.009 | |
219.01 | |
Crystalline Powder |
Chemical Identifiers
1006-41-3 | |
219.009 | |
RRKPMLZRLKTDQV-UHFFFAOYSA-N | |
1268169 | |
C1=CC(=C(C=C1F)Br)C(=O)O |
C7H4BrFO2 | |
MFCD00055370 | |
2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid | |
2-bromo-4-fluorobenzoic acid |
Safety and Handling
TSCA : No