Learn More
2-Bromo-4-fluorobenzoic acid, 98%, Thermo Scientific Chemicals
$100.42 - $281.23
Chemical Identifiers
CAS | 1006-41-3 |
---|---|
Molecular Formula | C7H4BrFO2 |
Molecular Weight (g/mol) | 219.009 |
MDL Number | MFCD00055370 |
InChI Key | RRKPMLZRLKTDQV-UHFFFAOYSA-N |
Synonym | 2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid |
PubChem CID | 1268169 |
IUPAC Name | 2-bromo-4-fluorobenzoic acid |
SMILES | C1=CC(=C(C=C1F)Br)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1932106
|
Thermo Scientific Chemicals
L1932106 |
5 g |
Each for $100.42
|
|
|||||
AAL1932114
|
Thermo Scientific Chemicals
L1932114 |
25 g |
Each for $281.23
|
|
|||||
Description
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.
Solubility
Soluble in water and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
1006-41-3 | |
219.009 | |
RRKPMLZRLKTDQV-UHFFFAOYSA-N | |
1268169 | |
C1=CC(=C(C=C1F)Br)C(=O)O |
C7H4BrFO2 | |
MFCD00055370 | |
2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid | |
2-bromo-4-fluorobenzoic acid |
Specifications
1006-41-3 | |
C7H4BrFO2 | |
5 g | |
2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid | |
RRKPMLZRLKTDQV-UHFFFAOYSA-N | |
2-bromo-4-fluorobenzoic acid | |
1268169 | |
98% |
167°C to 169°C | |
MFCD00055370 | |
2614292 | |
Soluble in water and methanol. | |
C1=CC(=C(C=C1F)Br)C(=O)O | |
219.009 | |
219.01 | |
2-Bromo-4-fluorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only