missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluorobenzenesulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ L1971306

 View more versions of this product

Catalog No. AAL1971306

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Description

2-Bromo-4-fluorobenzenesulfonyl chloride; 97%; Alfa Aesar; 5g; FW: 273.51; C6H3BrClFO2S; BP: 264 to 265; CAS: 351003-45-7

Applications
2-Bromo-4-fluorobenzenesulfonyl chloride is used as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
Description & Specifications

Specifications

2-Bromo-4-fluorobenzenesulfonyl chloride
97%
273.51
UN2920
5g
351003-45-7
C6H3BrClFO2S
MFCD03094149
264° to 265°C

Chemical Identifiers

351003-45-7
273.51
UN2920
C6H3BrClFO2S
MFCD03094149
2-Bromo-4-fluorobenzenesulfonyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.