Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluorobenzamide, 96%

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-fluorobenzamide
CAS 1006-40-2
Formula Weight 218.02
MDL Number MFCD06409122
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6249703

 
alfa aesar™
H6249703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6249706

 
alfa aesar™
H6249706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62497MD

 
alfa aesar™
H62497MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.02
CAS 1006-40-2
Quantity 1g
Assay 96%
MDL Number MFCD06409122
Chemical Name or Material 2-Bromo-4-fluorobenzamide
Physical Form Solid