missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluorobenzamide, 96%

Manufacturer:  Alfa Aesar™ H6249706

 View more versions of this product

Catalog No. AAH6249706



Specifications

Specifications

2-Bromo-4-fluorobenzamide
1006-40-2
MFCD06409122
IWODMEGZVDWKPK-UHFFFAOYSA-N
2-bromo-4-fluorobenzamide
20372234
Solid
5g
C7H5BrFNO
benzamide, 2-bromo-4-fluoro
C1=CC(=C(C=C1F)Br)C(=O)N
218.025
218.02
96%

Chemical Identifiers

1006-40-2
218.025
IWODMEGZVDWKPK-UHFFFAOYSA-N
20372234
C1=CC(=C(C=C1F)Br)C(=O)N
C7H5BrFNO
MFCD06409122
benzamide, 2-bromo-4-fluoro
2-bromo-4-fluorobenzamide
Documents