missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4'-fluoroacetophenone, 97%

Manufacturer:  Alfa Aesar™ A1531922

 View more versions of this product

Catalog No. AAA1531922



Description

Description

Applications
2-Bromo-4′-fluoroacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

2-Bromo-4'-fluoroacetophenone
150°C (12mmHg)
100g
C8H6BrFO
637863
ZJFWCELATJMDNO-UHFFFAOYSA-N
2-bromo-1-(4-fluorophenyl)ethanone
96749
97%
>110°C (230°F)
45°C to 49°C
403-29-2
MFCD00040830
2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone
C1=CC(=CC=C1C(=O)CBr)F
217.037
217.04

Chemical Identifiers

403-29-2
217.037
ZJFWCELATJMDNO-UHFFFAOYSA-N
96749
C1=CC(=CC=C1C(=O)CBr)F
C8H6BrFO
MFCD00040830
2-bromo-4'-fluoroacetophenone, 4-fluorophenacyl bromide, 2-bromo-1-4-fluorophenyl ethanone, 2-bromo-1-4-fluorophenyl ethan-1-one, p-fluorophenacyl bromide, 2'-bromo-4-fluoroacetophenone, ethanone, 2-bromo-1-4-fluorophenyl, bromo-4-fluoroacetophenone, 4-fluorophenacylbromide, 2-bromo-1-4-fluoro-phenyl-ethanone
2-bromo-1-(4-fluorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents