Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Bromo-4'-fluoroacetanilide, 98%

$28.78 - $272.45

Chemical Identifiers

Chemical Name or Material 2'-Bromo-4'-fluoroacetanilide
CAS 1009-22-9
Molecular Formula C8H7BrFNO
Formula Weight 232.05
MDL Number MFCD00061118
Beilstein 2803940
Synonym N-(2-Bromo-4-fluorophenyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2513306

 
alfa aesar™
B2513306
5g Each for $28.78

AAB2513322

 
alfa aesar™
B2513322
100g Each for $272.45
Description & Specifications

Specifications

Formula Weight 232.05
Chemical Name or Material 2'-Bromo-4'-fluoroacetanilide
Assay 98%
Molecular Formula C8H7BrFNO
MDL Number MFCD00061118
CAS 1009-22-9
Melting Point 116° to 119°C
Beilstein 2803940
Synonym N-(2-Bromo-4-fluorophenyl)acetamide
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.