missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluoro-6-nitroaniline, tech. 90%

Manufacturer:  Alfa Aesar™ H6184003

 View more versions of this product

Catalog No. AAH6184003



Description

Description

Applications
2-Bromo-4-fluoro-6-nitroaniline is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-Bromo-4-fluoro-6-nitroaniline
1g
C6H4BrFN2O2
UN2811
HCYDUPDSEDHSQB-UHFFFAOYSA-N
2-bromo-4-fluoro-6-nitroaniline
292004
Powder
Technical
Orange
10472-88-5
MFCD07779528
2-bromo-4-fluoro-6-nitrobenzenamine, 2-bromo-4-fluoro-6-nitrophenylamine, benzenamine, 2-bromo-4-fluoro-6-nitro, 2-amino-3-bromo-5-fluoronitrobenzene, pubchem16316, ksc495m4t, 4-fluoro-2-bromo-6-nitroaniline, 4-fluoro-2-nitro-6-bromoaniline, 6-bromo-4-fluoro-2-nitroaniline, 2-bromo-4-fluoro-6-nitroaniline
C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])F
235.012
235.01
90%

Chemical Identifiers

10472-88-5
235.012
HCYDUPDSEDHSQB-UHFFFAOYSA-N
292004
C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])F
C6H4BrFN2O2
MFCD07779528
2-bromo-4-fluoro-6-nitrobenzenamine, 2-bromo-4-fluoro-6-nitrophenylamine, benzenamine, 2-bromo-4-fluoro-6-nitro, 2-amino-3-bromo-5-fluoronitrobenzene, pubchem16316, ksc495m4t, 4-fluoro-2-bromo-6-nitroaniline, 4-fluoro-2-nitro-6-bromoaniline, 6-bromo-4-fluoro-2-nitroaniline, 2-bromo-4-fluoro-6-nitroaniline
2-bromo-4-fluoro-6-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents