Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluoro-5-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-fluoro-5-(trifluoromethyl)aniline
CAS 193090-60-7
Molecular Formula C7H4BrF4N
Formula Weight 258.01
MDL Number MFCD04973755
UN Number UN2811
Synonym 5-Amino-4-bromo-2-fluorobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2621106

 
alfa aesar™
H2621106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2621114

 
alfa aesar™
H2621114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.01
Chemical Name or Material 2-Bromo-4-fluoro-5-(trifluoromethyl)aniline
Assay 97%
Molecular Formula C7H4BrF4N
MDL Number MFCD04973755
CAS 193090-60-7
UN Number UN2811
Melting Point 24° to 29°C
Synonym 5-Amino-4-bromo-2-fluorobenzotrifluoride
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.