Learn More
2-Bromo-4-fluoro-5-nitrophenol, 97%, Thermo Scientific™
$115.63 - $446.60
Chemical Identifiers
CAS | 84478-87-5 |
---|---|
Molecular Formula | C6H3BrFNO3 |
Molecular Weight (g/mol) | 236.00 |
MDL Number | MFCD08460104 |
InChI Key | NVNFKCCUJKPLLT-UHFFFAOYSA-N |
Synonym | phenol, 2-bromo-4-fluoro-5-nitro, 2-bromo-4-fluoro-5-nitro-phenol, 4-bromo-2-fluoro-5-hydroxynitrobenzene, pubchem2837, ksc495s7j, 2-bromo-4-fluro-5-nitrophenol |
PubChem CID | 13268179 |
IUPAC Name | 2-bromo-4-fluoro-5-nitrophenol |
SMILES | OC1=C(Br)C=C(F)C(=C1)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6680103
|
Thermo Scientific Chemicals
H6680103 |
1 g |
Each for $115.63
|
|
|||||
AAH6680106
|
Thermo Scientific Chemicals
H6680106 |
5 g |
Each for $446.60
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
84478-87-5 | |
236.00 | |
NVNFKCCUJKPLLT-UHFFFAOYSA-N | |
13268179 | |
OC1=C(Br)C=C(F)C(=C1)[N+]([O-])=O |
C6H3BrFNO3 | |
MFCD08460104 | |
phenol, 2-bromo-4-fluoro-5-nitro, 2-bromo-4-fluoro-5-nitro-phenol, 4-bromo-2-fluoro-5-hydroxynitrobenzene, pubchem2837, ksc495s7j, 2-bromo-4-fluro-5-nitrophenol | |
2-bromo-4-fluoro-5-nitrophenol |
Specifications
84478-87-5 | |
Brown | |
MFCD08460104 | |
UN2811 | |
phenol, 2-bromo-4-fluoro-5-nitro, 2-bromo-4-fluoro-5-nitro-phenol, 4-bromo-2-fluoro-5-hydroxynitrobenzene, pubchem2837, ksc495s7j, 2-bromo-4-fluro-5-nitrophenol | |
OC1=C(Br)C=C(F)C(=C1)[N+]([O-])=O | |
236.00 | |
236 | |
Powder |
124°C to 127°C | |
C6H3BrFNO3 | |
1 g | |
15697238 | |
NVNFKCCUJKPLLT-UHFFFAOYSA-N | |
2-bromo-4-fluoro-5-nitrophenol | |
13268179 | |
97% | |
2-Bromo-4-fluoro-5-nitrophenol |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold