missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluoro-1-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ H6146622

 View more versions of this product

Catalog No. AAH6146622



Specifications

Specifications

2-Bromo-4-fluoro-1-nitrobenzene
41°C to 45°C
700-36-7
MFCD00792441
VGYVBEJDXIPSDL-UHFFFAOYSA-N
2-bromo-4-fluoro-1-nitrobenzene
2756993
Solid
Green to Yellow
100g
C6H3BrFNO2
1-bromo-5-fluoro-2-nitrobenzene, 2-bromo-4-fluoro-1-nitro-benzene, 2-bromo-4-fluoronitrobenzene, 3-bromo-1-fluoro-4-nitrobenzene, 4-fluoro-2-bromo nitrobenzene, 4-fluoro-2-bromonitrobenzene, acmc-1bkfy, benzene, 2-bromo-4-fluoro-1-nitro, pubchem4277, zlchem 479
C1=CC(=C(C=C1F)Br)[N+](=O)[O-]
219.997
220
98%

Chemical Identifiers

700-36-7
219.997
VGYVBEJDXIPSDL-UHFFFAOYSA-N
2756993
C1=CC(=C(C=C1F)Br)[N+](=O)[O-]
C6H3BrFNO2
MFCD00792441
1-bromo-5-fluoro-2-nitrobenzene, 2-bromo-4-fluoro-1-nitro-benzene, 2-bromo-4-fluoronitrobenzene, 3-bromo-1-fluoro-4-nitrobenzene, 4-fluoro-2-bromo nitrobenzene, 4-fluoro-2-bromonitrobenzene, acmc-1bkfy, benzene, 2-bromo-4-fluoro-1-nitro, pubchem4277, zlchem 479
2-bromo-4-fluoro-1-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents