missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-fluoro-1-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ H6146614

 View more versions of this product

Catalog No. AAH6146614



Specifications

Specifications

2-Bromo-4-fluoro-1-nitrobenzene
41°C to 45°C
700-36-7
MFCD00792441
VGYVBEJDXIPSDL-UHFFFAOYSA-N
2-bromo-4-fluoro-1-nitrobenzene
2756993
Solid
Green to Yellow
25g
C6H3BrFNO2
1-bromo-5-fluoro-2-nitrobenzene
C1=CC(=C(C=C1F)Br)[N+](=O)[O-]
219.997
220
98%

Chemical Identifiers

700-36-7
219.997
VGYVBEJDXIPSDL-UHFFFAOYSA-N
2756993
C1=CC(=C(C=C1F)Br)[N+](=O)[O-]
C6H3BrFNO2
MFCD00792441
1-bromo-5-fluoro-2-nitrobenzene
2-bromo-4-fluoro-1-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents