Learn More
2-Bromo-4'-(diethylamino)acetophenone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1333706
Description
2-Bromo-4'-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4′-(diethylamino)acetophenone, is used as a pharmaceutical intermediate. It is also used as a primary and secondary intermediate.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
2-Bromo-4'-(diethylamino)acetophenone | |
50°C to 52°C | |
MFCD00151798 | |
7921481 | |
It is insoluble in water. | |
CCN(CC)C1=CC=C(C=C1)C(=O)CBr | |
270.17 | |
270.18 |
207986-25-2 | |
C12H16BrNO | |
5 g | |
2-bromo-1-4-diethylamino phenyl ethanone, alpha-bromo-4-diethylamino acetophenone, 2-bromo-4'-diethylamino acetophenone, 2-bromo-1-4-diethylamino phenyl ethan-1-one, 4-diethylaminophenacyl bromide, n,n-diethyl-4-bromoacetyl aniline, 4-n,n-diethylamino phenacyl bromide, .alpha.-bromo-4-diethylaminoacetophenone, 2-bromo-1-4-diethylaminophenyl ethanone | |
ALTMCDJIFXTEPV-UHFFFAOYSA-N | |
2-bromo-1-[4-(diethylamino)phenyl]ethanone | |
2795030 | |
98% |
Chemical Identifiers
207986-25-2 | |
270.17 | |
ALTMCDJIFXTEPV-UHFFFAOYSA-N | |
2795030 | |
CCN(CC)C1=CC=C(C=C1)C(=O)CBr |
C12H16BrNO | |
MFCD00151798 | |
2-bromo-1-4-diethylamino phenyl ethanone, alpha-bromo-4-diethylamino acetophenone, 2-bromo-4'-diethylamino acetophenone, 2-bromo-1-4-diethylamino phenyl ethan-1-one, 4-diethylaminophenacyl bromide, n,n-diethyl-4-bromoacetyl aniline, 4-n,n-diethylamino phenacyl bromide, .alpha.-bromo-4-diethylaminoacetophenone, 2-bromo-1-4-diethylaminophenyl ethanone | |
2-bromo-1-[4-(diethylamino)phenyl]ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only