missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-4-chloroanisole, 98%, Thermo Scientific™
$87.41 - $87.41
Chemical Identifiers
CAS | 60633-25-2 |
---|---|
Molecular Formula | C7H6BrClO |
Molecular Weight (g/mol) | 221.48 |
MDL Number | MFCD00079705 |
InChI Key | YJEMGEBDXDPBSP-UHFFFAOYSA-N |
Synonym | 2-bromo-4-chloroanisole, 2-bromo-4-chloro-1-methoxy-benzene, benzene, 2-bromo-4-chloro-1-methoxy, 4-chloro-2-bromoanisole, pubchem2629, 2-bromo4-chloroanisole, 4-chloro-2-bromanisole, 2-bromo-4-chloro anisole, acmc-1b9j6, ksc490k3r |
PubChem CID | 521935 |
IUPAC Name | 2-bromo-4-chloro-1-methoxybenzene |
SMILES | COC1=C(C=C(C=C1)Cl)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC366560050
|
Thermo Scientific Chemicals
366560050 |
5 mL | Glass bottle |
Each for $87.41
|
|
Chemical Identifiers
60633-25-2 | |
221.48 | |
YJEMGEBDXDPBSP-UHFFFAOYSA-N | |
521935 | |
COC1=C(C=C(C=C1)Cl)Br |
C7H6BrClO | |
MFCD00079705 | |
2-bromo-4-chloroanisole, 2-bromo-4-chloro-1-methoxy-benzene, benzene, 2-bromo-4-chloro-1-methoxy, 4-chloro-2-bromoanisole, pubchem2629, 2-bromo4-chloroanisole, 4-chloro-2-bromanisole, 2-bromo-4-chloro anisole, acmc-1b9j6, ksc490k3r | |
2-bromo-4-chloro-1-methoxybenzene |
Specifications
60633-25-2 | |
100.0 | |
1.6250g/mL | |
Authentic | |
BrC6H3(Cl)OCH3 | |
Glass bottle | |
5 mL | |
1.625 | |
COC1=C(C=C(C=C1)Cl)Br | |
221.48 | |
221.48 | |
Liquid |
97.5 | |
Colorless to Yellow | |
97.5% min. (GC) | |
C7H6BrClO | |
MFCD00079705 | |
1.5850 to 1.5870 (20°C, 589nm) | |
2-bromo-4-chloroanisole, 2-bromo-4-chloro-1-methoxy-benzene, benzene, 2-bromo-4-chloro-1-methoxy, 4-chloro-2-bromoanisole, pubchem2629, 2-bromo4-chloroanisole, 4-chloro-2-bromanisole, 2-bromo-4-chloro anisole, acmc-1b9j6, ksc490k3r | |
YJEMGEBDXDPBSP-UHFFFAOYSA-N | |
2-bromo-4-chloro-1-methoxybenzene | |
521935 | |
98% | |
2-Bromo-4-chloroanisole, 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only