Please login to your online account to display your discounted pricing

2-Bromo-4-chloroaniline 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393470250

 View more versions of this product

Catalog No. AC393470250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 206.47
Quantity 25g
Color Pale brown
Melting Point 62° to 66°C
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(Br)NH2
CAS 873-38-1
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00041313
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2-Bromo-4-chloroaniline
Molecular Formula C6H5BrClN
Health Hazard 1 Danger

Specifications

Chemical Name or Material 2-Bromo-4-chloroaniline
CAS 873-38-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H5BrClN
Linear Formula ClC6H3(Br)NH2
Formula Weight 206.47
MDL Number MFCD00041313
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.

SDS & Safety Documents