Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-chloro-6-fluorophenol, 97%

$109.28 - $972.82

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-chloro-6-fluorophenol
CAS 886499-88-3
Molecular Formula C6H3BrClFO
Formula Weight 225.45
MDL Number MFCD04115955
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2677003

  SDS
alfa aesar™
H2677003
1g Each for $109.28

AAH2677006

  SDS
alfa aesar™
H2677006
5g Each for $323.24

AAH2677014

  SDS
alfa aesar™
H2677014
25g Each for $972.82
Description & Specifications

Specifications

Formula Weight 225.45
Chemical Name or Material 2-Bromo-4-chloro-6-fluorophenol
Assay 97%
Molecular Formula C6H3BrClFO
MDL Number MFCD04115955
CAS 886499-88-3
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.