Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4-chloro-6-fluoroanisole, 97%

$58.04 - $58.04

Chemical Identifiers

Chemical Name or Material 2-Bromo-4-chloro-6-fluoroanisole
Molecular Formula C7H5BrClFO
Formula Weight 239.47
MDL Number MFCD04115953
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2637503

  SDS
alfa aesar™
H2637503
1g Each for $58.04

AAH2637506

  SDS
alfa aesar™
H2637506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.47
Chemical Name or Material 2-Bromo-4-chloro-6-fluoroanisole
Assay 97%
Molecular Formula C7H5BrClFO
MDL Number MFCD04115953
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
An Acetanilide and Anisole derivative metabolite by liver. It is used to produce (5-fluoro-2-methoxy-phenyl)-(4-trifluoromethyl-phenyl)-methanone by reaction with 4-trifluoromethyl-benzonitrile. The reaction occurs with reagent Mg and solvent diethyl ether with other condition of heating for 2 hours. The yield is about 82%.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.