missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4-chloro-1-fluorobenzene, TCI America™

$105.42 - $316.25

Chemical Identifiers

CAS 1996-30-1
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00672934
InChI Key YFFUYGSLQXVHMB-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
PubChem CID 2773264
IUPAC Name 2-bromo-4-chloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29565G View Documents TCI America
B29565G
5g Each for $105.42
 
B295625G View Documents TCI America
B295625G
25g Each for $316.25
 
Specifications

Chemical Identifiers

1996-30-1
209.442
YFFUYGSLQXVHMB-UHFFFAOYSA-N
2773264
C1=CC(=C(C=C1Cl)Br)F
C6H3BrClF
MFCD00672934
3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
2-bromo-4-chloro-1-fluorobenzene

Specifications

90°C
5g
C6H3BrClF
3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
C1=CC(=C(C=C1Cl)Br)F
209.442
209.44
≥98.0% (GC)
Yellow
1996-30-1
MFCD00672934
YFFUYGSLQXVHMB-UHFFFAOYSA-N
2-bromo-4-chloro-1-fluorobenzene
2773264
Clear Liquid at 20°C
2-Bromo-4-chloro-1-fluorobenzene
Safety and Handling

Safety and Handling

TSCA : No