Please login to your online account to display your discounted pricing

2-Bromo-4,6-dinitroaniline 97%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC151221000

 View more versions of this product

Catalog No. AC151221000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0%
Formula Weight 262.01
Quantity 100g
Color Yellow to orange
EINECS Number 217-329-2
RTECS Number BW9303000
TSCA TSCA
Melting Point 148° to 153°C
CAS Min % 96.0
CAS Max % 100.0
MDL Number MFCD00007146
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.
Infrared Spectrum Authentic
Chemical Name or Material 2-Bromo-4, 6-dinitroaniline
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6H4BrN3O4
CAS 1817-73-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Bromo-4, 6-dinitroaniline
Name Note 97%
CAS 1817-73-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H4BrN3O4
Formula Weight 262.01
MDL Number MFCD00007146
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.

SDS & Safety Documents