Please login to your online account to display your discounted pricing

2-Bromo-4,6-dinitroaniline 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromo-4, 6-dinitroaniline
Name Note 97%
CAS 1817-73-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H4BrN3O4
Formula Weight 262.01
MDL Number MFCD00007146
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC151221000

  SDS
thermo fisher scientific
AC151221000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 262.01
Assay ≥96.0%
Quantity 100g
Color Yellow to orange
RTECS Number BW9303000
EINECS Number 217-329-2
TSCA TSCA
Melting Point 148° to 153°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.
Infrared Spectrum Authentic
MDL Number MFCD00007146
CAS 1817-73-8
Name Note 97%
Molecular Formula C6H4BrN3O4
Assay Percent Range 97%
Chemical Name or Material 2-Bromo-4, 6-dinitroaniline
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable solid.