Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,6-dinitroaniline, 97%

$23.05 - $55.76

Chemical Identifiers

Chemical Name or Material 2-Bromo-4,6-dinitroaniline
CAS 1817-73-8
Molecular Formula C6H4BrN3O4
Formula Weight 262.02
MDL Number MFCD00007146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2290514

  SDS
alfa aesar™
B2290514
25g Each for $23.05

AAB2290522

  SDS
alfa aesar™
B2290522
100g Each for $55.76
Description & Specifications

Specifications

Formula Weight 262.02
CAS 1817-73-8
Quantity 25g
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Chemical Name or Material 2-Bromo-4,6-dinitroaniline
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 151° to 153°C
TSCA TSCA
Assay 97%
MDL Number MFCD00007146
Health Hazard 1 Warning
Molecular Formula C6H4BrN3O4
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.