Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,6-difluorophenol, 98+%

$89.80 - $331.49

Chemical Identifiers

Chemical Name or Material 2-Bromo-4,6-difluorophenol
CAS 98130-56-4
Molecular Formula C6H3BrF2O
Formula Weight 208.99
MDL Number MFCD00070752
Beilstein 3245985
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2518606

  SDS
alfa aesar™
B2518606
5g Each for $89.80

AAB2518614

  SDS
alfa aesar™
B2518614
25g Each for $331.49
Description & Specifications

Specifications

Formula Weight 208.99
Flash Point 105°C (221°F)
Beilstein 3245985
MDL Number MFCD00070752
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
CAS 98130-56-4
Boiling Point 53°C (1mmHg)
Assay ≥98%
Health Hazard 1 Warning
Molecular Formula C6H3BrF2O
Chemical Name or Material 2-Bromo-4,6-difluorophenol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.