Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,6-difluoroanisole, 97%

$39.03 - $485.18

Chemical Identifiers

Chemical Name or Material 2-Bromo-4,6-difluoroanisole
CAS 202865-59-6
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD00070754
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2619903

  SDS
alfa aesar™
H2619903
1g Each for $39.03

AAH2619906

  SDS
alfa aesar™
H2619906
5g Each for $150.36

AAH2619914

  SDS
alfa aesar™
H2619914
25g Each for $485.18
Description & Specifications

Specifications

Formula Weight 223.02
CAS 202865-59-6
Quantity 1g
MDL Number MFCD00070754
Assay 97%
Molecular Formula C7H5BrF2O
Chemical Name or Material 2-Bromo-4,6-difluoroanisole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.