missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4,6-dichloroaniline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC399600250

 View more versions of this product

Catalog No. AC399600250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-4, 6-dichloroaniline
80.0°C to 81.0°C
697-86-9
100.0
C6H4BrCl2N
MFCD00040936
DTPADCOGQUOGHT-UHFFFAOYSA-N
2-bromo-4,6-dichloroaniline
2756901
98%
Glass bottle
25g
97.5
97.5% min. (GC)
Cl2C6H2(Br)NH2
2,4-dichloro-6-bromoaniline, 2-bromanyl-4,6-bis chloranyl aniline, 2-bromo-4,6-dichloro-aniline, 2-bromo-4,6-dichloroaniline, 6-bromo-2,4-dichloroaniline, 6-bromo-2,4-dichlorophenylamine, acmc-209ob3, benzenamine,2-bromo-4,6-dichloro, pubchem3605
C1=C(C=C(C(=C1Cl)N)Br)Cl
240.91
240.91

Chemical Identifiers

697-86-9
240.91
DTPADCOGQUOGHT-UHFFFAOYSA-N
2756901
C1=C(C=C(C(=C1Cl)N)Br)Cl
C6H4BrCl2N
MFCD00040936
2,4-dichloro-6-bromoaniline, 2-bromanyl-4,6-bis chloranyl aniline, 2-bromo-4,6-dichloro-aniline, 2-bromo-4,6-dichloroaniline, 6-bromo-2,4-dichloroaniline, 6-bromo-2,4-dichlorophenylamine, acmc-209ob3, benzenamine,2-bromo-4,6-dichloro, pubchem3605
2-bromo-4,6-dichloroaniline
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning

SDS Documents