missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-4,6-dichloroaniline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 399600050
Specifications
2-Bromo-4, 6-dichloroaniline | |
97.5 | |
97.5% min. (GC) | |
C6H4BrCl2N | |
MFCD00040936 | |
pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline | |
C1=C(C=C(C(=C1Cl)N)Br)Cl | |
240.91 | |
240.91 |
697-86-9 | |
100.0 | |
Glass bottle | |
Cl2C6H2(Br)NH2 | |
5g | |
DTPADCOGQUOGHT-UHFFFAOYSA-N | |
2-bromo-4,6-dichloroaniline | |
2756901 | |
98% |
Chemical Identifiers
697-86-9 | |
240.91 | |
DTPADCOGQUOGHT-UHFFFAOYSA-N | |
2756901 | |
C1=C(C=C(C(=C1Cl)N)Br)Cl |
C6H4BrCl2N | |
MFCD00040936 | |
pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline | |
2-bromo-4,6-dichloroaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
RUO â Research Use Only