Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,5-methylenedioxycinnamic acid, 97%

$54.39 - $54.39

Chemical Identifiers

Chemical Name or Material 2-Bromo-4,5-methylenedioxycinnamic acid
CAS 27452-00-2
Molecular Formula C10H7BrO4
Formula Weight 271.07
MDL Number MFCD00017599
Synonym 3-(6-Bromo-1,3-benzodioxol-5-yl)propenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1820803

  SDS
alfa aesar™
A1820803
1g Each for $54.39
Description & Specifications

Specifications

Formula Weight 271.07
Chemical Name or Material 2-Bromo-4,5-methylenedioxycinnamic acid
Melting Point ≈255°C (dec.)
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00017599
Assay 97%
Health Hazard 1 Warning
Molecular Formula C10H7BrO4
CAS 27452-00-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 3-(6-Bromo-1,3-benzodioxol-5-yl)propenoic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.