Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,5-dimethoxycinnamic acid, 97%

$59.51 - $681.05

Chemical Identifiers

Chemical Name or Material 2-Bromo-4,5-dimethoxycinnamic acid
CAS 151539-52-5
Molecular Formula C11H11BrO4
Formula Weight 287.12
MDL Number MFCD00017614
Beilstein 3142451
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1686206

  SDS
alfa aesar™
A1686206
5g Each for $59.51

AAA1686214

  SDS
alfa aesar™
A1686214
25g Each for $216.97

AAA1686222

  SDS
alfa aesar™
A1686222
100g Each for $681.05
Description & Specifications

Specifications

Formula Weight 287.12
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 151539-52-5
Quantity 5g
MDL Number MFCD00017614
Melting Point 253° to 256°C
Health Hazard 1 Warning
Chemical Name or Material 2-Bromo-4,5-dimethoxycinnamic acid
Beilstein 3142451
Assay 97%
Molecular Formula C11H11BrO4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.