missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-4,5-dimethoxybenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC395090010

 View more versions of this product

Catalog No. AC395090010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Bromo-4, 5-dimethoxybenzoic acid
96.0
Authentic
Powder
97%
(CH3O)2C6H2(Br)CO2H
MFCD00017542
185° to 189°C
6286-46-0
100.0
Glass bottle
≥96.0%
C9H9BrO4
261.07
White to cream or yellow
1g

Chemical Identifiers

6286-46-0
100.0
C9H9BrO4
261.07
2-Bromo-4, 5-dimethoxybenzoic acid
96.0
97%
(CH3O)2C6H2(Br)CO2H
MFCD00017542
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.