Please login to your online account to display your discounted pricing

2-Bromo-4,5-dimethoxybenzoic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromo-4, 5-dimethoxybenzoic acid
CAS 6286-46-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9BrO4
Linear Formula (CH3O)2C6H2(Br)CO2H
Formula Weight 261.07
MDL Number MFCD00017542
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC395090010

  SDS
thermo fisher scientific
AC395090010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC395090050

  SDS
thermo fisher scientific
AC395090050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 261.07
Quantity 1g
Color White to cream or yellow
Melting Point 185° to 189°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Bromo-4, 5-dimethoxybenzoic acid
Packaging Glass bottle
Molecular Formula C9H9BrO4
CAS 6286-46-0
Assay Percent Range 97%
Linear Formula (CH3O)2C6H2(Br)CO2H
MDL Number MFCD00017542
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.