Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,5-difluorotoluene, 98%

$143.92 - $360.19

Chemical Identifiers

Chemical Name or Material 2-Bromo-4,5-difluorotoluene
CAS 875664-38-3
Molecular Formula C7H5BrF2
Formula Weight 207.02
MDL Number MFCD07777160
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2614303

 
alfa aesar™
H2614303
1g Each for $360.19

AAH26143MD

 
alfa aesar™
H26143MD
250mg Each for $143.92
Description & Specifications

Specifications

Formula Weight 207.02
Chemical Name or Material 2-Bromo-4,5-difluorotoluene
Assay 98%
Molecular Formula C7H5BrF2
MDL Number MFCD07777160
CAS 875664-38-3
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.