missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,5-difluorophenylacetic acid, 98%

Manufacturer:  Alfa Aesar™ H63902MD

 View more versions of this product

Catalog No. AAH63902MD



Description

Description

Applications
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Bromo-4,5-difluorophenylacetic acid
250mg
C8H5BrF2O2
2-2-bromo-4,5-difluorophenyl acetic acid, 2-2-bromo-4,5-difluorophenyl aceticacid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylacetic acid, 2-bromo-4,5-difluorophenylaceticacid, acmc-209qsj, benzeneacetic acid, 2-bromo-4,5-difluoro, pubchem2702
FQSLXVRCDYUESO-UHFFFAOYSA-N
2-(2-bromo-4,5-difluorophenyl)acetic acid
3732964
Solid
White
883502-07-6
MFCD04973423
Slightly soluble in water.
C1=C(C(=CC(=C1F)F)Br)CC(=O)O
251.027
251.03
98%

Chemical Identifiers

883502-07-6
251.027
FQSLXVRCDYUESO-UHFFFAOYSA-N
3732964
C1=C(C(=CC(=C1F)F)Br)CC(=O)O
C8H5BrF2O2
MFCD04973423
2-2-bromo-4,5-difluorophenyl acetic acid, 2-2-bromo-4,5-difluorophenyl aceticacid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylacetic acid, 2-bromo-4,5-difluorophenylaceticacid, acmc-209qsj, benzeneacetic acid, 2-bromo-4,5-difluoro, pubchem2702
2-(2-bromo-4,5-difluorophenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents