missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,5-difluorophenylacetic acid, 98%

Manufacturer:  Alfa Aesar™ H6390203

 View more versions of this product

Catalog No. AAH6390203



Description

Description

Applications
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Bromo-4,5-difluorophenylacetic acid
1g
C8H5BrF2O2
2-bromo-4,5-difluorophenylacetic acid, 2-2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylaceticacid, pubchem2702, acmc-209qsj, 2-2-bromo-4,5-difluorophenyl aceticacid, benzeneacetic acid, 2-bromo-4,5-difluoro, 2-bromo-4,5-difluorophenylacetic acid
C1=C(C(=CC(=C1F)F)Br)CC(=O)O
251.027
251.03
98%
White
883502-07-6
MFCD04973423
FQSLXVRCDYUESO-UHFFFAOYSA-N
2-(2-bromo-4,5-difluorophenyl)acetic acid
3732964
Solid

Chemical Identifiers

883502-07-6
251.027
FQSLXVRCDYUESO-UHFFFAOYSA-N
3732964
C1=C(C(=CC(=C1F)F)Br)CC(=O)O
C8H5BrF2O2
MFCD04973423
2-bromo-4,5-difluorophenylacetic acid, 2-2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenyl acetic acid, 2-bromo-4,5-difluorophenylaceticacid, pubchem2702, acmc-209qsj, 2-2-bromo-4,5-difluorophenyl aceticacid, benzeneacetic acid, 2-bromo-4,5-difluoro, 2-bromo-4,5-difluorophenylacetic acid
2-(2-bromo-4,5-difluorophenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents