Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-4,5-difluoroanisole, 98%

$452.03 - $452.03

Chemical Identifiers

Chemical Name or Material 2-Bromo-4,5-difluoroanisole
CAS 202865-58-5
Molecular Formula C7H5BrF2O
Formula Weight 223.02
MDL Number MFCD00070753
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2179209

  SDS
alfa aesar™
B2179209
10g Each for $452.03
Description & Specifications

Specifications

Formula Weight 223.02
Quantity 10g
Health Hazard 1 Warning
MDL Number MFCD00070753
CAS 202865-58-5
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 98%
Chemical Name or Material 2-Bromo-4,5-difluoroanisole
Molecular Formula C7H5BrF2O

Applications
It is employed as a intermediate for pharmaceuticals.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.